Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599651 Supplier Invitrogen™ Supplier No. PA599651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MCT12.

This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc16a12
Gene Accession No. Q6ZSM3
Gene Alias 3230401A21; AW210596; CJMG; Creatine transporter 2; CRT2; LOW QUALITY PROTEIN: monocarboxylate transporter 12; MCT 12; MCT12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; RGD1311468; SLC16A12; solute carrier family 16 (monocarboxylic acid transporters), member 12; solute carrier family 16 member 12; solute carrier family 16, member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Symbols Slc16a12
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC16A12(Accession Q6ZSM3), corresponding to amino acid residues I444-T486.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387700
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.