Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC16A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106681
Description
Antibody detects endogenous levels of total MOT13.
Monocarboxylates, such as lactate and pyruvate, play an integral role in cellular metabolism. Lactic acid is produced in large quantities as a result of glycolysis, which provides the majority of ATP to cells under normal physiological conditions. However, accumulation of lactic acid leads to a decrease in intracellular pH and, thus, to a cessation of glycolysis. In order for glycolysis to continue at a high rate, lactic acid must be transported out of the cell. This transport process is carried out by a family of monocarboxylate transporters (MCTs), which function as proton symports and are stereoselective for L-lactate. MCT13 (Monocarboxylate transporter 13), also known as SLC16A13 (Solute carrier family 16 member 13), is a 426 amino acid multi-pass membrane protein that belongs to the MCT transport family. Functioning as a proton-linked monocarboxylate transporter, MCT13 catalyzes the rapid transports of molecules, such as lactate, across the plasma membrane.
Specifications
SLC16A13 | |
Polyclonal | |
Unconjugated | |
SLC16A13 | |
1700007D07Rik; MCT 13; MCT13; monocarboxylate transporter; Monocarboxylate transporter 13; monocarboxylic acid transporter 13; MOT13; Slc16a13; solute carrier family 16 (monocarboxylic acid transporters), member 13; solute carrier family 16 member 13; solute carrier family 16, member 13; solute carrier family 16, member 13 (monocarboxylic acid transporter 13) | |
Rabbit | |
Affinity chromatography | |
RUO | |
201232, 287451, 69309 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q66HE2, Q7RTY0, Q8CE94 | |
SLC16A13 | |
A synthesized peptide derived from human SLC16A13(Accession Q7RTY0), corresponding to amino acid residues P392-D426. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction