Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559207 Supplier Invitrogen™ Supplier No. PA559207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YTSHEDIGYD FEDGPKDKKT LKPHPNIDGG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Proton-linked monocarboxylate transporter. May catalyze the transport of monocarboxylates across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A14
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC16A14
Gene Accession No. Q7RTX9
Gene Alias 1110004H10Rik; MCT 14; MCT14; monocarboxylate transporter; Monocarboxylate transporter 14; monocarboxylic acid transporter 14; MOT14; Slc16a14; solute carrier family 16 (monocarboxylic acid transporters), member 14; solute carrier family 16 member 14; solute carrier family 16, member 14 (monocarboxylic acid transporter 14)
Gene Symbols SLC16A14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC16A14. Recombinant protein control fragment (Product #RP-96174).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151473
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.