Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106682 Supplier Invitrogen™ Supplier No. PA5106682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MOT14.

Proton-linked monocarboxylate transporter. May catalyze the transport of monocarboxylates across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC16A14
Gene Accession No. Q7RTX9, Q8K1C7
Gene Alias 1110004H10Rik; MCT 14; MCT14; monocarboxylate transporter; Monocarboxylate transporter 14; monocarboxylic acid transporter 14; MOT14; Slc16a14; solute carrier family 16 (monocarboxylic acid transporters), member 14; solute carrier family 16 member 14; solute carrier family 16, member 14 (monocarboxylic acid transporter 14)
Gene Symbols SLC16A14
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC16A14(Accession Q7RTX9), corresponding to amino acid residues F130-K180.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151473, 71781
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.