Learn More
Invitrogen™ SLC16A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596251
Description
Immunogen sequence: MALQSQASEE AKGPWQEADQ EQQEPVGSPE PESEPEPEPE PEPVPVPPPE PQPEPQPLPD PAPLPELEFE SERVHEPEPT PTVETRGTAR GFQPPEGGFG; Positive Samples: LO2, HT-29, 293T, Mouse heart, Mouse kidney, Mouse liver, Rat heart, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.
Monocarboxylates, such as lactate and pyruvate, play an integral role in cellular metabolism. Lactic acid is produced in large quantities as a result of glycolysis, which provides the majority of ATP to cells under normal physiological conditions. However, accumulation of lactic acid leads to a decrease in intracellular pH and cessation of glycolysis. In order for glycolysis to continue at a high rate, lactic acid must be transported out of the cell. This transport process is carried out by a family of monocarboxylate transporters (MCTs), which function as proton symports and are stereoselective for L-lactate. The MCT family consists of at least eight members, MCT 1-8, which contain between 10-12 transmembrane-helical (TM) domains, with the amino and carboxy termini located in the cytoplasm. Defects in the gene encoding for MCT8, SLC16A2, can cause monocarboxylate transporter 8 deficiency (MCT8 deficiency), a defect in cellular hormone transport causing a severe form of X-linked psychomotor retardation and abnormal thyroid levels.
Specifications
SLC16A2 | |
Polyclonal | |
Unconjugated | |
Slc16a2 | |
AHDS; AW105741; DXS128; DXS128E; MCT 7; MCT 8; MCT7; MCT8; monocarboxylate transporter; monocarboxylate transporter 7; Monocarboxylate transporter 8; Monocarboxylate transporter 8 (MCT 8) (X-linked PEST-containing transporter); MRX22; SLC16A2; solute carrier family 16 (monocarboxylic acid transporters), member 2; solute carrier family 16 member 2; solute carrier family 16, member 2 (monocarboxylic acid transporter 8); solute carrier family 16, member 2 (thyroid hormone transporter); X-linked PEST-containing transporter; XPCT | |
Rabbit | |
Affinity chromatography | |
RUO | |
20502, 259248, 6567 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.78 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O70324, P36021, Q8K1P8 | |
Slc16a2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SLC16A2 (NP_0065082). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.