Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC16A3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP181251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP181251 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP181251 Supplier Novus Biologicals Supplier No. NBP181251
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

SLC16A3 Polyclonal specifically detects SLC16A3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen SLC16A3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot Reported in scientific literature (PMID:33212210), Simple Western 1:250, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen Reactivity reported in (PMID:24692237), Immunoblotting Reactivity reported in (PMID: 28978054).
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O15427
Gene Alias MCT 3, MCT3MCT 4, MCT4, MCT4MGC138472, MGC138474, Monocarboxylate transporter 3, monocarboxylate transporter 4, solute carrier family 16 (monocarboxylic acid transporters), member 3, Solute carrier family 16 member 3, solute carrier family 16, member 3 (monocarboxylic acid transporter 4)
Gene Symbols SLC16A3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9123
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.