Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC16A6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159881
Description
SLC16A6 Polyclonal specifically detects SLC16A6 in Human samples. It is validated for Western Blot.Specifications
SLC16A6 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
MCT6MCT 6, MCT7MCT 7, Monocarboxylate transporter 6, monocarboxylate transporter 7, solute carrier family 16 (monocarboxylic acid transporters), member 6, Solute carrier family 16 member 6, solute carrier family 16, member 6 (monocarboxylic acid transporter 7) | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 92%; Canine: 85%; Rat: 85%; Bovine: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
O15403 | |
SLC16A6 | |
Synthetic peptides corresponding to SLC16A6(solute carrier family 16, member 6 (monocarboxylic acid transporter 7)) The peptide sequence was selected from the middle region of SLC16A6. Peptide sequence ILKEKSFICYALFGLFATLGFFAPSLYIIPLGISLGIDQDRAAFLLSTMA The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
9120 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction