Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569267 Supplier Invitrogen™ Supplier No. PA569267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.

Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC16A6
Gene Accession No. O15403
Gene Alias AW743111; breast cancer protein BCY5; EGK_08910; ESTM12; MCT 6; MCT 7; MCT6; MCT7; Mct7B; monocarboxylate transporter; Monocarboxylate transporter 6; monocarboxylate transporter 7; monocarboxylate transporter 7B; monocarboxylate transporter 7-like; MOT7; Slc16a6; solute carrier family 16 (monocarboxylic acid transporters), member 6; solute carrier family 16 member 6; solute carrier family 16, member 6; solute carrier family 16, member 6 (monocarboxylic acid transporter 7)
Gene Symbols SLC16A6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC16A6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9120
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.