Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598155
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598155 Supplier Invitrogen™ Supplier No. PA598155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC16A8 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SLC16A8 is proton-linked monocarboxylate transporter. It catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC16A8
Gene Accession No. O35308, O95907
Gene Alias LOW QUALITY PROTEIN: monocarboxylate transporter 3; MCT 3; MCT3; monocarboxylate transporter; monocarboxylate transporter 3; monocarboxylate transporter 3; LOW QUALITY PROTEIN: monocarboxylate transporter 3; preferentially expressed in RPE cells and choroid plexus epithelium; proton-coupled monocarboxylate transporter 3; REMP; Slc16a8; solute carrier 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 (monocarboxylate transporter), member 8; solute carrier family 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 member 8; solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Gene Symbols SLC16A8
Host Species Rabbit
Immunogen Recombinant Human Monocarboxylate transporter 3 protein (432-504AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23539, 57274
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.