Learn More
Invitrogen™ SLC16A8 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SLC16A8.
Specifications
Specifications
| Antigen | SLC16A8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SLC16A8 |
| Gene Accession No. | O35308, O70461, O95907 |
| Gene Alias | LOW QUALITY PROTEIN: monocarboxylate transporter 3; MCT 3; MCT3; monocarboxylate transporter; monocarboxylate transporter 3; monocarboxylate transporter 3; LOW QUALITY PROTEIN: monocarboxylate transporter 3; preferentially expressed in RPE cells and choroid plexus epithelium; proton-coupled monocarboxylate transporter 3; REMP; Slc16a8; solute carrier 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 (monocarboxylate transporter), member 8; solute carrier family 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 member 8; solute carrier family 16, member 8 (monocarboxylic acid transporter 3) |
| Gene Symbols | SLC16A8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.