Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561924 Supplier Invitrogen™ Supplier No. PA561924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQSSDCPLPK KIAPEDLPDK YSIYNEKGKN LEENINILDK SYSSEEKCRI TLANGDWKQD SLLHK Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.

Predicted to enable monocarboxylic acid transmembrane transporter activity. Involved in urate metabolic process. Predicted to be integral component of plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC16A9
Gene Accession No. Q7RTY1
Gene Alias 1200003C15Rik; 4930425B13Rik; AI552636; C10orf36; I79_016926; MCT 9; MCT9; monocarboxylate transporter; Monocarboxylate transporter 9; monocarboxylic acid transporter 9; Slc16a9; solute carrier family 16 (monocarboxylic acid transporters), member 9; solute carrier family 16 member 9; solute carrier family 16, member 9; solute carrier family 16, member 9 (monocarboxylic acid transporter 9)
Gene Symbols SLC16A9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC16A9. Recombinant protein control fragment (Product #RP-97135).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.