Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC17A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545587 Supplier Invitrogen™ Supplier No. PA545587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VHCCNVIITA QRACLNLTMV VMVNSTDPHG LPNTSTKKLL DNIKNPMYNW Sequence homology: Cow: 85%; Dog: 83%; Horse: 100%; Human: 100%; Pig: 77%.

SLC17A1 (Solute Carrier Family 17 Member 1) is a Protein Coding gene. Diseases associated with SLC17A1 include Gout and Hyperuricemia. Among its related pathways are Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds and Uricosurics Pathway, Pharmacodynamics. Gene Ontology (GO) annotations related to this gene include symporter activity and phosphate ion transmembrane transporter activity. An important paralog of this gene is SLC17A2. Also, it is involved in cotransport of sodium and phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc17a1
Gene Accession No. Q14916
Gene Alias Na(+)/PI cotransporter 1; Na/Pi-4; Napi1; Napi-1; Npt1; NPT-1; OATV1; organic anion transporter OATv1; renal Na(+)-dependent phosphate cotransporter 1; Renal sodium-dependent phosphate transport protein 1; renal sodium-phosphate transport protein 1; Slc17a1; sodium phosphate transporter; sodium/phosphate cotransporter 1; sodium/phosphate type I cotransporter; sodium-dependent phosphate transport protein 1; sodium-dependent phosphate/anion cotransporter; solute carrier family 17 (organic anion transporter), member 1; solute carrier family 17 (sodium phosphate), member 1; solute carrier family 17 (sodium/hydrogen exchanger), member 1; solute carrier family 17 (vesicular glutamate transporter), member 1; solute carrier family 17 member 1; solute carrier family 17 vesicular glutamate transporter), member 1
Gene Symbols Slc17a1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC17A1 (aa 26-75).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6568
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.