Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC17A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568625 Supplier Invitrogen™ Supplier No. PA568625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

SLC17A2 gene ontology annotations related to this gene include transmembrane transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC17A2
Gene Accession No. Q5SZA1
Gene Alias C730032N17Rik; Na(+)/PI cotransporter 3; Npt3; SLC17A2; sodium phosphate transporter 3; sodium/phosphate cotransporter 3; sodium-dependent phosphate transport protein 3; sodium-dependent phosphate transport protein 3-like; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (vesicular glutamate transporter), member 2; solute carrier family 17 member 2; Solute carrier family 17 member 3; solute carrier family 17, member 2; solute carrier family 34 (sodium phosphate), member 1; Unknown (protein for MGC:134070)
Gene Symbols SLC17A2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC17A2 (aa 281-330).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218103
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.