Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC17A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542138 Supplier Invitrogen™ Supplier No. PA542138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FVVIYDDPVS YPWISTSEKE YIISSLKQQV GSSKQPLPIK AMLRSLPIWS Sequence homology: Cow: 85%; Horse: 85%; Human: 100%.

SLC17A3 may be involved in actively transporting phosphate into cells via Na(+) cotransport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC17A3
Gene Accession No. O00476
Gene Alias GOUT4; Na(+)/PI cotransporter 4; NPT4; SLC17A3; sodium/phosphate cotransporter 4; Sodium-dependent phosphate transport protein 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; solute carrier family 17 member 3; UAQTL4
Gene Symbols SLC17A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC17A3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10786
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.