Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC17A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565955 Supplier Invitrogen™ Supplier No. PA565955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDDPVNHPFIS AGEKRYIVCS LAQQDCSPGW SLPIRA Highest antigen sequence identity to the following orthologs - mouse 81%, rat 84%.

As a Na/PO4 cotransporter, SLC17A4 may be important to the regulation of Li transport and its therapeutic effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC17A4
Gene Accession No. Q9Y2C5
Gene Alias 9130214H05Rik; KAIA2138; Na/PO4 cotransporter; probable small intestine urate exporter; putative small intestine sodium-dependent phosphate transport protein; Slc17a4; solute carrier family 17 (sodium phosphate), member 4; solute carrier family 17 member 4; solute carrier family 17, member 4
Gene Symbols SLC17A4
Host Species Rabbit
Immunogen Recombinant Human SLC17A4. Recombinant protein control fragment (Product #RP-106644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.