Learn More
Invitrogen™ SLC17A4 Polyclonal Antibody

Description
Peptide sequence: YFCEYWLFYT IMAYTPTYIS SVLQANLRDS GILSALPFVV GCICIILGGL Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | SLC17A4 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SLC17A4 |
| Gene Accession No. | Q9Y2C5 |
| Gene Alias | 9130214H05Rik; KAIA2138; Na/PO4 cotransporter; probable small intestine urate exporter; putative small intestine sodium-dependent phosphate transport protein; Slc17a4; solute carrier family 17 (sodium phosphate), member 4; solute carrier family 17 member 4; solute carrier family 17, member 4 |
| Gene Symbols | SLC17A4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.