Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC17A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542456 Supplier Invitrogen™ Supplier No. PA542456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LFTPIAADLG VGPLIVLRAL EGLGEGVTFP AMHAMWSSWA PPLERSKLLS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%; Zebrafish: 93%.

This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC17A5
Gene Accession No. Q9NRA2
Gene Alias 4631416G20Rik; 4732491M05; AST; FLJ22227; FLJ23268; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; hp59; ISSD; membrane glycoprotein HP59; Membrane glycoprotein SP55; NSD; proton-coupled sialic acid transporter; sb:cb809; SD; sialic acid storage disease; sialin; SIASD; SLC17A5; SLD; Sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; solute carrier family 17 (anion/sugar transporter), member 5; solute carrier family 17 member 5; SP55; Vesicular H(+)/Aspartate-glutamate cotransporter; zgc:153077
Gene Symbols SLC17A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC17A5 (aa 151-200).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26503
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.