Learn More
SLC17A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111910
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KGEVQSW ALSDHHG Homology: Rat - 13/14 amino acid residues identical; human - 12/14 amino acid residues identical.
This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.
Specifications
SLC17A5 | |
Polyclonal | |
Unconjugated | |
SLC17A5 | |
4631416G20Rik; 4732491M05; AST; BOS_9478; FLJ22227; FLJ23268; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; ISSD; Membrane glycoprotein HP59; Membrane glycoprotein SP55; NSD; sb:cb809; SD; Sialin; SIASD; SLC17A5; SLD; Sodium/sialic acid cotransporter; Solute carrier family 17 member 5; SP55; Vesicular H(+)/Aspartate-glutamate cotransporter; zgc:153077 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
235504, 26503, 363103 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q8BN82, Q9NRA2 | |
SLC17A5 | |
Peptide corresponding to amino acid residues 479-492 of mouse SLC17A5 (Intracellular, C-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.