Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC17A5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9934 Supplier Bioss Supplier No. BS24067R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.

Specifications

Antigen SLC17A5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene SLC17A5
Gene Accession No. Q9NRA2
Gene Alias 4631416G20Rik; 4732491M05; AST; BOS_9478; FLJ22227; FLJ23268; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; ISSD; Membrane glycoprotein HP59; Membrane glycoprotein SP55; NSD; proton-coupled sialic acid transporter; sb:cb809; SD; sialic acid storage disease; Sialin; SIASD; SLC17A5; SLD; Sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; solute carrier family 17 (anion/sugar transporter), member 5; Solute carrier family 17 member 5; SP55; Vesicular H(+)/Aspartate-glutamate cotransporter; zgc:153077
Gene Symbols SLC17A5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse SLC17A5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26503
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.