Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC17A6A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572190
Description
SLC17A6A Polyclonal Antibody for Western Blot
Mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells.
Specifications
SLC17A6A | |
Polyclonal | |
Unconjugated | |
slc17a6a | |
slc17a6a; slc17a6l; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6 like; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6a; solute carrier family 17 (vesicular glutamate transporter), member 6a; solute carrier family 17 member 6 B; solute carrier family 17 member 6a; solute carrier family 17 member 6-A; Vesicular glutamate transporter 2.2; vesicular glutamate transporter 2-B; vglut2.2; vglut22; vglut2b; wu:fj12c01 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
494492 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.66 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5W8I7 | |
slc17a6a | |
KLH conjugated synthetic peptide located within amino acid positions 557-587 amino acids of DANRE slc17a6a. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction