Learn More
SLC17A9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111874
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)SQKVQES ERAFTYS Homology: Human - 13/14 amino acid residues identical; rat - 12/14 amino acid residues identical.
Involved in vesicular storage and exocytosis of ATP. May accumulate ATP and other nucleotides in secretory vesicles such as adrenal chromaffin granules and synaptic vesicles.
Specifications
| SLC17A9 | |
| Polyclonal | |
| Unconjugated | |
| Slc17a9 | |
| 1700019H03Rik; BC019537; C20orf59; POROK8; RGD1311940; SLC17A9; Solute carrier family 17 member 9; VNUT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 228993, 362287, 63910 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q8VCL5, Q9BYT1 | |
| Slc17a9 | |
| Peptide corresponding to amino acid residues 155-168 of mouse VNUT (Intracellular, 2nd loop) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.