Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC18A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564912 Supplier Invitrogen™ Supplier No. PA564912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTFLYDMEFKE VNSSLHLGHA GSSPHALASP AFSTIFSFFN NNTVAVEESV PSGIAWMNDT ASTIPPPATE AISAHKNNCL QGTGFLEEEI Highest antigen sequence identity to the following orthologs - mouse 64%, rat 65%.

SLC18A1 (Solute Carrier Family 18 Member A1) is a Protein Coding gene. Diseases associated with SLC18A1 include Barbiturate Abuse and Adrenal Neuroblastoma. Among its related pathways are Transport of inorganic cations/anions and amino acids/oligopeptides and Synaptic vesicle pathway. Gene Ontology (GO) annotations related to this gene include monoamine transmembrane transporter activity and xenobiotic transmembrane transporter activity. An important paralog of this gene is SLC18A2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC18A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC18A1
Gene Accession No. P54219
Gene Alias CGAT; VMAT
Gene Symbols SLC18A1
Host Species Rabbit
Immunogen Recombinant Human SLC18A1. Recombinant protein control fragment (Product #RP-88660).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.