Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC18B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565574 Supplier Invitrogen™ Supplier No. PA565574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEALGDLEGPR APGGDDPAGS AGETPGWLSR Highest antigen sequence identity to the following orthologs - mouse 46%, rat 44%.

This gene encodes a protein, which has high sequence similarity to rat, xenopus and zebrafish proteins. The protein function is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC18B1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC18B1
Gene Accession No. Q6NT16
Gene Alias 1110021L09Rik; C6orf192; dJ55C23.6; MFS-type transporter C6orf192; MFS-type transporter C6orf192 homolog; MFS-type transporter SLC18B1; RGD1306962; SLC18B1; Solute carrier family 18 member B1; solute carrier family 18, subfamily B, member 1
Gene Symbols SLC18B1
Host Species Rabbit
Immunogen Recombinant Human SLC18B1. Recombinant protein control fragment (Product #RP-104880).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.