Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC19A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588778 Supplier Invitrogen™ Supplier No. PA588778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDGLRHCQRG HHPRQPPAQG LRSAAEEKAA QALSVQDKGL GGLQPAQSPP LSPEDSLGAV GPASLEQRQS DPYLAQAPAP QAAEFLSPVT TPSPCTLCSA QASGPEAADE TCPQLAVHPP GVSKLGLQCL PSDGVQNVNQ; Positive Samples: HeLa, U-87MG; Cellular Location: Membrane, Multi-pass membrane protein.

Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC19A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC19A1
Gene Accession No. P41440
Gene Alias AI323572; CHMD; FLOT1; folate transporter 1; FOLT; IFC1; IFC-1; Intestinal folate carrier 1; intestinal folate carrier protein; methotrexate carrier 1; methotrexate carrier 2; methotrexate carrier 5; methotrexate carrier 6; MTX1; MTX-1; Placental folate transporter; reduced folate carrier 1; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; REFC; RFC; RFC1; RFC-1; RFT-1; SLC19A1; solute carrier family 19 (folate transporter), member 1; solute carrier family 19 (sodium/hydrogen exchanger), member 1; solute carrier family 19 member 1; solute carrier family 19, member 1
Gene Symbols SLC19A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 452-591 of human SLC19A1 (NP_9192311).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6573
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.