Learn More
Invitrogen™ SLC19A2 Polyclonal Antibody

Description
Immunogen sequence: RECWFLPTAL LCAYGFFASL RPSEPFLTPY LLGPDKNLTE REEPKPDRLL VLKVLWNDFL MCYSSR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 62%.
Specifications
Specifications
| Antigen | SLC19A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC19A2 |
| Gene Accession No. | O60779 |
| Gene Alias | AV276020; AW322295; DDA1; high affinity thiamine transporter; reduced folate carrier protein (RFC) like; Slc19a2; solute carrier family 19 (thiamine transporter), member 2; solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; THMD1; THT1; THTR1; ThTr-1; TRMA |
| Gene Symbols | SLC19A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.