Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC19A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578332 Supplier Invitrogen™ Supplier No. PA578332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, mouse liver Predicted Reactivity: Rat (82%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SLC19A2 encodes a thiamin transporter protein. Mutations in this gene cause thiamin-responsive megaloblastic anemia syndrome (TRMA), which is an autosomal recessive disorder characterized by diabetes mellitus, megaloblastic anemia and sensorineural deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC19A2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SLC19A2
Gene Accession No. O60779, Q9EQN9
Gene Alias AV276020; AW322295; DDA1; high affinity thiamine transporter; reduced folate carrier protein (RFC) like; Slc19a2; solute carrier family 19 (thiamine transporter), member 2; solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; THMD1; THT1; THTR1; ThTr-1; TRMA
Gene Symbols SLC19A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human SLC19A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10560, 116914
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.