Learn More
SLC19A2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA552255
Description
This gene encodes the thiamin transporter protein. Mutations in this gene cause thiamin-responsive megaloblastic anemia syndrome (TRMA), which is an autosomal recessive disorder characterized by diabetes mellitus, megaloblastic anemia and sensorineural deafness.
Immunogen sequence: LFFHHIPSTC QRVNGIKVQN GGIVTDTPAS NHLPGWEDIE SKIPLNMEEP PVEEPEPKPD RLLVLKVLWN DFLMCYSSRHighest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
Specifications
SLC19A2 | |
Polyclonal | |
Unconjugated | |
SLC19A2 | |
TC1, THT1, TRMA, THMD1, THTR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10560 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.1mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60779 | |
SLC19A2 | |
Recombinant protein corresponding to Human SLC19A2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only