Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC19A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100333 Supplier Invitrogen™ Supplier No. PA5100333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC19A3.

SLC19A3 encodes a ubiquitously expressed transmembrane thiamine transporter that lacks folate transport activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC19A3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC19A3
Gene Accession No. Q99PL8, Q9BZV2
Gene Alias A230084E24Rik; AI788884; BBGD; Slc19a3; solute carrier family 19 (sodium/hydrogen exchanger), member 3; solute carrier family 19 (thiamine transporter), member 3; solute carrier family 19 member 3; solute carrier family 19, member 3; thiamine transporter 2; THMD2; THTR2; thTr-2
Gene Symbols SLC19A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC19A3(Accession Q9BZV2), corresponding to amino acid residues T220-K270.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316559, 80704, 80721
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.