Learn More
Invitrogen™ SLC19A3 Polyclonal Antibody

Description
SLC19A3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SLC19A3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SLC19A3 |
| Gene Accession No. | Q9BZV2 |
| Gene Alias | A230084E24Rik; AI788884; BBGD; Slc19a3; solute carrier family 19 (sodium/hydrogen exchanger), member 3; solute carrier family 19 (thiamine transporter), member 3; solute carrier family 19 member 3; solute carrier family 19, member 3; thiamine transporter 2; THMD2; THTR2; thTr-2 |
| Gene Symbols | SLC19A3 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.