Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC19A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598872
Description
SLC19A3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
SLC19A3 encodes a ubiquitously expressed transmembrane thiamine transporter that lacks folate transport activity.
Specifications
SLC19A3 | |
Polyclonal | |
Unconjugated | |
SLC19A3 | |
A230084E24Rik; AI788884; BBGD; Slc19a3; solute carrier family 19 (sodium/hydrogen exchanger), member 3; solute carrier family 19 (thiamine transporter), member 3; solute carrier family 19 member 3; solute carrier family 19, member 3; thiamine transporter 2; THMD2; THTR2; thTr-2 | |
Rabbit | |
Protein G | |
RUO | |
80704 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
3.55 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9BZV2 | |
SLC19A3 | |
Recombinant Human Thiamine transporter 2 protein (191-282AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction