Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC1A1, Mouse anti-Human, PerCP, Clone: S180-41, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB18296
Specifications
SLC1A1 | |
Monoclonal | |
PerCP | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
EAAC1/EAAT3 | |
Mouse | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
S180-41 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1 | |
SLC1A1 | |
Recombinant protein corresponding to full length human SLC1A1. | |
RUO | |
6505 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction