Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A4/ASCT1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0010 Supplier Bethyl Laboratories, Inc Supplier No. A305280A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.

Specifications

Antigen SLC1A4/ASCT1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SLC1A4
Gene Accession No. P43007
Gene Alias ASCT1, SATT, ASCT-1, alanine/serine/cysteine/threonine transporter 1, glutamate/neutral amino acid transporter, neutral amino acid transporter A, solute carrier family 1 member 4
Gene Symbols SLC1A4
Host Species Rabbit
Immunogen Between 482 and 532
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6509
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.