Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC1A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576693 Supplier Invitrogen™ Supplier No. PA576693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC1A4
Gene Accession No. O35874, P43007
Gene Alias Alanine/serine/cysteine/threonine transporter 1; ASCT1; ASCT-1; AW045657; glutamate/neutral amino acid transporter; neutral amino acid transporter A; neutral amino acid transporter ASCT1; OTTHUMP00000159933; OTTHUMP00000235138; SATT; si:zc101n13.5; Slc1a4; slc1a4 protein; sodium-dependent neutral amino acid transporter ASCT1; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; solute carrier family 1 member 4; SPATCCM; zgc:92817
Gene Symbols SLC1A4
Host Species Rabbit
Immunogen Recombinant full length Human SLC1A4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305540, 55963, 6509
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.