Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5120853
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120853 Supplier Thermo Scientific Supplier No. PA5120853
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, BT-474, H460. The target is usually found in the following locations: Melanosome, Membrane, Multi-pass membrane protein.

SLC1A4 Polyclonal antibody specifically detects SLC1A4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC1A4
Gene Accession No. O35874, P43007
Gene Alias Alanine/serine/cysteine/threonine transporter 1; ASCT1; ASCT-1; AW045657; glutamate/neutral amino acid transporter; neutral amino acid transporter A; neutral amino acid transporter ASCT1; OTTHUMP00000159933; OTTHUMP00000235138; SATT; si:zc101n13.5; Slc1a4; slc1a4 protein; sodium-dependent neutral amino acid transporter ASCT1; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; solute carrier family 1 member 4; SPATCCM; zgc:92817
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human SLC1A4 (P43007).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305540, 55963, 6509
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.