Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC1A7 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281802
Description
SLC1A7 Polyclonal specifically detects SLC1A7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC1A7 | |
Polyclonal | |
Unconjugated | |
PBS | |
SLC1A7 | |
SLC1A7 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human SLC1A7. The immunogen is located within amino acids 460 - 510 of SLC1A7. Amino Acid Squence: RKDFARDTGTEKL | |
Affinity Purified | |
RUO | |
6512 | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
AAAT, EAAT5, excitatory amino acid transporter 5 (retinal glutamate transporter), excitatory amino acid transporter 5a long, retinal glutamate transporter, solute carrier family 1 (glutamate transporter), member 7 | |
Rabbit | |
52 kDa | |
0.1 mg | |
Primary | |
SLC1A7 antibody is human and mouse reactive. Multiple isoforms of SLC1A7 are known to exist. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction