Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC1A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561887 Supplier Invitrogen™ Supplier No. PA561887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLVEATFKQY RTKTTPVVKS PKVAPEEAPP RRILIYGVQE ENGSHVQNFA LDLTPPPEVV YKSEPGTSDG M Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

This gene belongs to the sodium:dicarboxylate (SDF) symporter (TC 2.A.23) family, SLC1A7 subfamily. As transportor of L-glutamate; the L-glutamate uptake is sodium- and voltage-dependent and chloride-independent. Its associated chloride conductance may participate in visual processing.It interacts with the PDZ domains of DLG4. Expressed primarily in retina. Detectable in liver, heart, muscle and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc1a7
Gene Accession No. O00341
Gene Alias A930031E15Rik; AAAT; EAAT5; Excitatory amino acid transporter 5; excitatory amino acid transporter 5 (retinal glutamate transporter); retinal glutamate transporter; SLC1A7; solute carrier family 1 (glutamate transporter), member 7; solute carrier family 1 member 7
Gene Symbols Slc1a7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC1A7. Recombinant protein control fragment (Product #RP-98525).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.