Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572847
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572847 Supplier Thermo Scientific Supplier No. PA572847
Add to Cart
Add to Cart

SLC1A7 Polyclonal Antibody

SLC1A7 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Slc1a7
Gene Accession No. O00341, Q8JZR4
Gene Alias A930031E15Rik; AAAT; EAAT5; Excitatory amino acid transporter 5; excitatory amino acid transporter 5 (retinal glutamate transporter); retinal glutamate transporter; SLC1A7; solute carrier family 1 (glutamate transporter), member 7; solute carrier family 1 member 7
Gene Symbols Slc1a7
Host Species Rabbit
Immunogen SLC1A7 antibody for a 14 amino acid peptide near the carboxy terminus of human SLC1A7. The immunogen is within amino acids 460 - 510 of SLC1A7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242607, 6512
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.