Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC20A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23826625UL
Description
SLC20A1 Polyclonal specifically detects SLC20A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC20A1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q8WUM9 | |
SLC20A1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KIEREIKCSPSESPLMEKKNSLKEDHEETKLSVGDIENKHPVSEVGPATVPLQAVVEERTVSFKLGDLEEAPERERLPSVDL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ41426, Gibbon ape leukemia virus receptor 1, Glvr-1, GLVR1DKFZp686J2397, Leukemia virus receptor 1 homolog, Phosphate transporter 1, PiT-1PIT1, sodium-dependent phosphate transporter 1, solute carrier family 20 (phosphate transporter), member 1, Solute carrier family 20 member 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
6574 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction