Learn More
Invitrogen™ SLC20A1 Polyclonal Antibody

Description
Recommended positive controls: H1299, mouse brain. Predicted reactivity: Mouse (95%), Rat (94%), Cat (95%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SLC20A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | SLC20A1 |
| Gene Accession No. | Q61609, Q8WUM9 |
| Gene Alias | AI607883; DKFZp686J2397; FLJ41426; Gibbon ape leukemia virus receptor 1; GLVR1; Glvr-1; leukemia virus receptor 1 homolog; Phosphate transporter 1; PIT1; piT-1; RPHO-1; Slc20a1; Sodium-dependent phosphate transporter 1; Sodium-dependent phosphate transporter 1-like protein; solute carrier family 20 (phosphate transporter), member 1; solute carrier family 20 member 1; solute carrier family 20, member 1 |
| Gene Symbols | SLC20A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.