Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC20A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598650
Description
SLC20A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Retrovirus receptors allow infection of human and murine cells by various retroviruses. The receptors that have been identified at the molecular level include CD4 for human immunodeficiency virus, Rec1 for murine ecotropic virus, and GLVR1 for gibbon ape leukemia virus. These 3 proteins show no homology to one another at the DNA or protein level. GLVR1 is a sodium-dependent phosphate symporter.
Specifications
SLC20A1 | |
Polyclonal | |
Unconjugated | |
SLC20A1 | |
AI607883; DKFZp686J2397; FLJ41426; Gibbon ape leukemia virus receptor 1; GLVR1; Glvr-1; leukemia virus receptor 1 homolog; Phosphate transporter 1; PIT1; piT-1; RPHO-1; Slc20a1; Sodium-dependent phosphate transporter 1; Sodium-dependent phosphate transporter 1-like protein; solute carrier family 20 (phosphate transporter), member 1; solute carrier family 20 member 1; solute carrier family 20, member 1 | |
Rabbit | |
Protein G | |
RUO | |
6574 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8WUM9 | |
SLC20A1 | |
Recombinant Human Sodium-dependent phosphate transporter 1 protein (263-458AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction