Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC20A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528184 Supplier Invitrogen™ Supplier No. PA528184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Unboiled U87-MG, Boiled A431, Unboiled A431. Predicted reactivity: Mouse (85%), Rat (87%), Cat (90%), Rhesus Monkey (99%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC20A2
Applications Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SLC20A2
Gene Accession No. Q08357
Gene Alias Ab1-188; fi23g11; Gibbon ape leukemia virus receptor 2; Glvr2; GLVR-2; hPit2; IBGC1; IBGC3; MLVAR; MolPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; phosphate transporter 2; PIT2; PIT-2; RAM1; RAM-1; receptor for amphitropic viruses 1; receptor for amphotropic viruses 1; slc20a2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; solute carrier family 20 member 2; solute carrier family 20, member 2; type III sodium-dependent phosphate transporter; wu:fi23g11; zgc:152990
Gene Symbols SLC20A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PiT2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.