Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC20A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121031 Supplier Invitrogen™ Supplier No. PA5121031
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BT-474, HepG2, Mouse liver, Mouse kidney, Rat liver. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein.

This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC20A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC20A2
Gene Accession No. Q08357, Q63488, Q80UP8
Gene Alias Ab1-188; fi23g11; Gibbon ape leukemia virus receptor 2; Glvr2; GLVR-2; hPit2; IBGC1; IBGC3; MLVAR; MolPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; phosphate transporter 2; PIT2; PIT-2; RAM1; RAM-1; receptor for amphitropic viruses 1; receptor for amphotropic viruses 1; slc20a2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; solute carrier family 20 member 2; solute carrier family 20, member 2; type III sodium-dependent phosphate transporter; wu:fi23g11; zgc:152990
Gene Symbols SLC20A2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20516, 29502, 6575
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.