Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A1 Monoclonal Antibody (2C5)
GREENER_CHOICE

Catalog No. PIMA515730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515730 Supplier Invitrogen™ Supplier No. MA515730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15730 targets SLC22A1 in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15730 immunogen is purified recombinant fragment of human SLC22A1 expressed in E. Coli. MA5-15730 detects SLC22A1 which has a predicted molecular weight of approximately 61.2kDa.

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2C5
Concentration Concentration not determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene SLC22A1
Gene Accession No. O15245
Gene Alias 1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK
Gene Symbols SLC22A1
Host Species Mouse
Immunogen Purified recombinant fragment of human SLC22A1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.