Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580014
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580014 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580014 Supplier Invitrogen™ Supplier No. PA580014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA Cell, A549 Cell.

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SLC22A1
Gene Accession No. O15245
Gene Alias 1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK
Gene Symbols SLC22A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human SLC22A1 (533-550aa RKAKPKENTIYLKVQTSE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6580
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.