Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555285
Description
Immunogen sequence: RWLIIKGKPD QALQELRKVA RINGHKEAKN LTIEVLMSSV KEEVASAKEP RSVLDLFCVP V Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 52%.
SLC22A11 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A11 is an integral membrane protein and is found mainly in the kidney and in the placenta, where it may act to prevent potentially harmful organic anions from reaching the fetus.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and is found mainly in the kidney and in the placenta, where it may act to prevent potentially harmful organic anions from reaching the fetus. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
SLC22A11 | |
Polyclonal | |
Unconjugated | |
SLC22A11 | |
hOAT4; MGC34282; OAT4; Organic anion transporter 4; SLC22A11; solute carrier family 22 (organic anion/cation transporter), member 11; solute carrier family 22 (organic anion/urate transporter), member 11; solute carrier family 22 member 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55867 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NSA0 | |
SLC22A11 | |
Recombinant protein corresponding to Human SLC22A11. Recombinant protein control fragment (Product #RP-88589). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction