Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596820 Supplier Invitrogen™ Supplier No. PA596820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSAAIPGHRC WTHMLDNGSA VSTNMTPKAL LTISIPPGPN QGPHQCRRFR QPQWQLLDPN ATATSWSEAD TEPCVDGWVY DRSVFTSTIV AKWDLVCSSQ G Positive Samples: SW620, BT-474, HepG2, 293T, Mouse kidney, Mouse liver, Rat kidney, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.

SLC22A11 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A11 is an integral membrane protein and is found mainly in the kidney and in the placenta, where it may act to prevent potentially harmful organic anions from reaching the fetus.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and is found mainly in the kidney and in the placenta, where it may act to prevent potentially harmful organic anions from reaching the fetus. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC22A11
Gene Accession No. Q9NSA0
Gene Alias hOAT4; MGC34282; OAT4; Organic anion transporter 4; SLC22A11; solute carrier family 22 (organic anion/cation transporter), member 11; solute carrier family 22 (organic anion/urate transporter), member 11; solute carrier family 22 member 11
Gene Symbols SLC22A11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-140 of human SLC22A11 (NP_0609541).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55867
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.