Learn More
Invitrogen™ SLC22A12 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SLC22A12 protein.
Specifications
Specifications
| Antigen | SLC22A12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | SLC22A12 |
| Gene Accession No. | Q96S37 |
| Gene Alias | AI987855; OAT4L; OATL4; Organic anion transporter 4-like protein; renal-specific transporter; RST; Slc22a12; Slc22al2; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; solute carrier family 22 member 12; UNQ6453/PRO34004; URAT1; Urate anion exchanger 1; urate transporter 1 |
| Gene Symbols | SLC22A12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.