Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576559
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SLC22A12 | |
Polyclonal | |
Unconjugated | |
SLC22A12 | |
AI987855; OAT4L; OATL4; Organic anion transporter 4-like protein; renal-specific transporter; RST; Slc22a12; Slc22al2; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; solute carrier family 22 member 12; UNQ6453/PRO34004; URAT1; Urate anion exchanger 1; urate transporter 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116085, 20521, 365398 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3ZAV1, Q8CFZ5, Q96S37 | |
SLC22A12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-150 of human SLC22A12. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction