Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106386
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106386 Supplier Invitrogen™ Supplier No. PA5106386
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total URAT1.

SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC22A12
Gene Accession No. Q3ZAV1, Q8CFZ5, Q96S37
Gene Alias AI987855; OAT4L; OATL4; Organic anion transporter 4-like protein; renal-specific transporter; RST; Slc22a12; Slc22al2; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; solute carrier family 22 member 12; UNQ6453/PRO34004; URAT1; Urate anion exchanger 1; urate transporter 1
Gene Symbols SLC22A12
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC22A12(Accession Q96S37), corresponding to amino acid residues D55-A105.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116085, 20521, 365398
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.