Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557336 Supplier Invitrogen™ Supplier No. PA557336
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PETHGQGLKD TLQDLELGPH PRSPKSVPSE KETEAKGRTS SPGVAFVSST Y Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 65%.

SLC22A13 is a member of the organic-cation transporter family. SLC22A13 is a transmembrane protein involved in the transport of small molecules. This protein can function to mediate urate uptake and is a high affinity nicotinate exchanger in the kidneys and the intestine.This gene encodes a member of the organic-cation transporter family. It is located in a gene cluster with another member of the family, organic cation transporter like 4. The encoded protein is a transmembrane protein which is thought to transport small molecules and since this protein is conserved among several species, it is suggested to have a fundamental role in mammalian systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC22A13
Gene Accession No. Q9Y226
Gene Alias AI648912; OAT10; OCTL1; OCTL3; ORCTL3; ORCTL-3; Organic cation transporter-like 3; organic cationic transporter-like 3; organic-cation transporter like 3; Slc22a13; solute carrier family 22 (organic anion transporter), member 13; solute carrier family 22 (organic anion/urate transporter), member 13; solute carrier family 22 (organic cation transporter), member 13; solute carrier family 22 member 13; solute carrier family 22, member 13
Gene Symbols SLC22A13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC22A13. Recombinant protein control fragment (Product #RP-96458).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9390
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.